(Phenylthio)acetic acid 95%
Supplier: Apollo Scientific
OR916733-100G
OR916733-25G
APOSOR916733-100GEA
192
GBP
APOSOR916733-100G
APOSOR916733-25G
(Phenylthio)acetic acid 95%
(Phenylthio)acetic acid
Formula:
C₈H₈O₂S MW: 168,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004355 CAS Number: 103-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...