Heptamethyleneimine 98%
Supplier: Apollo Scientific
Synonyms:
Azacyclooctane, Azocane
OR916642-1G
OR916642-25G
OR916642-5G
OR916642-250MG
OR916642-10G
APOSOR916642-1GEA
72.2
GBP
APOSOR916642-1G
APOSOR916642-25G
APOSOR916642-5G
APOSOR916642-250MG
APOSOR916642-10G
Heptamethyleneimine 98%
Heptamethyleneimine
Formula:
C₇H₁₅N MW: 113,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003270 CAS Number: 1121-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...