1-Bromo-4-(tert-butylsulphanyl)benzene 95%
Supplier: Apollo Scientific
|
Warning
|
OR914606-1G
OR914606-5G
APOSOR914606-1GEA
165
GBP
APOSOR914606-1G
APOSOR914606-5G
1-Bromo-4-(tert-butylsulphanyl)benzene 95%
1-Bromo-4-(tert-butylsulphanyl)benzene
Formula:
C₁₀H₁₃BrS MW: 245,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973729 CAS Number: 25752-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...