Ethyl pyrrolidinoacetate 98%

Supplier: Apollo Scientific
Warning

OR914405-100G OR914405-25G OR914405-5G
APOSOR914405-100GEA 649 GBP
APOSOR914405-100G APOSOR914405-25G APOSOR914405-5G
Ethyl pyrrolidinoacetate 98%
Ethyl pyrrolidinoacetate
Formula: C₈H₁₅NO₂
MW: 157,213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012085
CAS Number: 22041-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR