Methyl-3-(4-hydroxyphenyl)propionate 98%

Supplier: Apollo Scientific

Synonyms: 3-(4-Hydroxyphenyl)propionic Acid Methyl Ester, Methyl 3-(4-hydroxyphenyl)propanoate, Methyl 3-(4-hydroxyphenyl)propionate, 3-(4-Hydroxy-phenyl)-propionic acid-OMe

OR914364-100G OR914364-25G
APOSOR914364-100GEA 80 GBP
APOSOR914364-100G APOSOR914364-25G
Methyl-3-(4-hydroxyphenyl)propionate 98%
Methyl-3-(4-hydroxyphenyl)propionate
Formula: C₁₀H₁₂O₃
MW: 180,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071577
CAS Number: 5597-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR