4-Imidazol-1-ylmethylphenylamine 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-[(1H-Imidazol-1-yl)methyl]aniline
OR9141-1G
OR9141-5G
APOSOR9141-1GEA
74.8
GBP
APOSOR9141-1G
APOSOR9141-5G
4-Imidazol-1-ylmethylphenylamine 95%
4-Imidazol-1-ylmethylphenylamine
Formula:
C₁₀H₁₁N₃ MW: 173,2144 g/mol Melting Pt: 128…130 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06660785 CAS Number: 56643-85-7 UN: 2923 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...