2-Methyl-3-(methylthio)furan 95%
Supplier: Apollo Scientific
OR913430-10G
OR913430-25G
OR913430-5G
APOSOR913430-10GEA
257
GBP
APOSOR913430-10G
APOSOR913430-25G
APOSOR913430-5G
2-Methyl-3-(methylthio)furan 95%
2-Methyl-3-(methylthio)furan
Formula:
C₆H₈OS MW: 128,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03701635 CAS Number: 63012-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...