(6-Methoxy-pyridin-2-yl)methanol 95%

Supplier: Apollo Scientific

Synonyms: (6-Methoxypyridin-2-yl)methanol

OR913313-1G OR913313-250MG
APOSOR913313-1GEA 124 GBP
APOSOR913313-1G APOSOR913313-250MG
(6-Methoxy-pyridin-2-yl)methanol 95%
(6-Methoxy-pyridin-2-yl)methanol
Formula: C₇H₉NO₂
MW: 139,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235128
CAS Number: 63071-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR