2-(4-Methylphenyl)piperazine 95%

Supplier: Apollo Scientific
Warning

OR913098-1G OR913098-250MG
APOSOR913098-1GEA 466 GBP
APOSOR913098-1G APOSOR913098-250MG
2-(4-Methylphenyl)piperazine 95%
2-(4-Methylphenyl)piperazine
Formula: C₁₁H₁₆N₂
Storage Temperature: Ambient
CAS Number: 65709-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR