5-Methyl-1H-indazole-3-carboxylic acid 95%
Supplier: Apollo Scientific
OR913075-1G
OR913075-250MG
APOSOR913075-1GEA
82.6
GBP
APOSOR913075-1G
APOSOR913075-250MG
5-Methyl-1H-indazole-3-carboxylic acid 95%
5-Methyl-1H-indazole-3-carboxylic acid
Formula:
C9H8N2O2 MW: 176,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05663986 CAS Number: 1201-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...