2-Amino-3-chlorobenzonitrile 98%

Supplier: Apollo Scientific

OR912867-25G OR912867-5G
APOSOR912867-25GEA 190 GBP
APOSOR912867-25G APOSOR912867-5G
2-Amino-3-chlorobenzonitrile 98%
2-Amino-3-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425881
CAS Number: 53312-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR