2-Methyl-3-(propyldithio)furan 95%
Supplier: Apollo Scientific
Synonyms:
2-Methyl-3-(propyldisulphanyl)furan, 2-Methyl-3-furyl Propyl Disulphide
OR912698-10G
OR912698-1G
OR912698-5G
APOSOR912698-10GEA
55.1
GBP
APOSOR912698-10G
APOSOR912698-1G
APOSOR912698-5G
2-Methyl-3-(propyldithio)furan 95%
2-Methyl-3-(propyldithio)furan
Formula:
C₈H₁₂OS₂ MW: 188,30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01729816 CAS Number: 61197-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...