2-(3-Chlorophenyl)piperazine 95%

Supplier: Apollo Scientific

OR912572-1G OR912572-250MG
APOSOR912572-1GEA 486 GBP
APOSOR912572-1G APOSOR912572-250MG
2-(3-Chlorophenyl)piperazine 95%
2-(3-Chlorophenyl)piperazine
Formula: C₁₀H₁₃ClN₂
Storage Temperature: Ambient
CAS Number: 52385-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR