2,3,4,5-Tetrahydro-1H-benzo[b]azepine 95%
Supplier: Apollo Scientific
OR912503-1G
OR912503-5G
APOSOR912503-1GEA
69.5
GBP
APOSOR912503-1G
APOSOR912503-5G
2,3,4,5-Tetrahydro-1H-benzo[b]azepine 95%
2,3,4,5-Tetrahydro-1H-benzo[b]azepine
Formula:
C₁₀H₁₃N MW: 147,2200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00272363 CAS Number: 1701-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...