Ethyl indole-3-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Indole-3-carboxylic acid ethyl ester, Ethyl 1H-indole-3-carboxylate

OR912467-1G OR912467-5G
APOSOR912467-1GEA 48.6 GBP
APOSOR912467-1G APOSOR912467-5G
Ethyl indole-3-carboxylate 95%
Ethyl indole-3-carboxylate
Formula: C₁₁H₁₁NO₂
MW: 189,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00228454
CAS Number: 776-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR