5-Chloropyrimidin-2(1H)-one 98%
Supplier: Apollo Scientific
Synonyms:
5-Chloro-2-hydroxy-pyrimidine·HCl
OR912364-1G
OR912364-25G
OR912364-5G
APOSOR912364-1GEA
112
GBP
APOSOR912364-1G
APOSOR912364-25G
APOSOR912364-5G
5-Chloropyrimidin-2(1H)-one 98%
5-Chloropyrimidin-2(1H)-one
Formula:
C4H3ClN2O Storage Temperature: Ambient |
MDL Number:
MFCD09250871 CAS Number: 54326-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...