2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran 97%
Supplier: Apollo Scientific
Synonyms:
2,2-Dimethyl-7-hydroxycoumaran, 2,2-Dimethyl-2,3-dihydrobenzofuran-7-ol
OR912320-100G
OR912320-500G
APOSOR912320-100GEA
165
GBP
APOSOR912320-100G
APOSOR912320-500G
2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran 97%
2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran
Formula:
C₁₀H₁₂O₂ MW: 164,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00075382 CAS Number: 1563-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...