4-Chloro-α,α-dimethylphenylacetic acid 98%

Supplier: Apollo Scientific

Synonyms: 2-(4-Chlorophenyl)-2-methylpropanoic acid

OR912236-10G OR912236-25G
APOSOR912236-10GEA 171 GBP
APOSOR912236-10G APOSOR912236-25G
4-Chloro-α,α-dimethylphenylacetic acid 98%
4-Chloro-alpha,alpha-dimethylphenylacetic acid
Formula: C₁₀H₁₁ClO₂
MW: 198,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044814
CAS Number: 6258-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR