Methyl-2-amino-5-methoxybenzoate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 2-amino-5-methoxybenzoate

OR912063-10G OR912063-1G OR912063-5G
APOSOR912063-10GEA 840 GBP
APOSOR912063-10G APOSOR912063-1G APOSOR912063-5G
Methyl-2-amino-5-methoxybenzoate 98%
Methyl 2-amino-5-methoxybenzoate
Formula: C₉H₁₁NO₃
MW: 181,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038482
CAS Number: 2475-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR