4-(4-Oxocyclohexyl)benzonitrile ≥98%

Supplier: Apollo Scientific

OR912053-10G OR912053-25G OR912053-5G
APOSOR912053-10GEA 94.4 GBP
APOSOR912053-10G APOSOR912053-25G APOSOR912053-5G
4-(4-Oxocyclohexyl)benzonitrile ≥98%
4-(4-Oxocyclohexyl)benzonitrile
Formula: C₁₃H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD06410874
CAS Number: 73204-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR