4-Phenoxypyridine 95%
Supplier: Apollo Scientific
OR911925-1G
OR911925-25G
OR911925-5G
APOSOR911925-1GEA
98.3
GBP
APOSOR911925-1G
APOSOR911925-25G
APOSOR911925-5G
4-Phenoxypyridine 95%
4-Phenoxypyridine
Formula:
C₁₁H₉NO MW: 171,2000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00235157 CAS Number: 4783-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...