2,6-Quinolinediol 98%
Supplier: Apollo Scientific
Synonyms:
6-Hydroxyquinolin-2(1H)-one, 2,6-Dihydroxyquinoline
OR911797-100G
OR911797-25G
APOSOR911797-100GEA
185
GBP
APOSOR911797-100G
APOSOR911797-25G
2,6-Quinolinediol 98%
2,6-Quinolinediol
Formula:
C₉H₇NO₂ MW: 161,1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00239446 CAS Number: 19315-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...