5-Chlorovaleric acid 97%
Supplier: Apollo Scientific
OR911386-100G
OR911386-500G
APOSOR911386-100GEA
54.3
GBP
APOSOR911386-100G
APOSOR911386-500G
5-Chlorovaleric acid 97%
5-Chlorovaleric acid
Formula:
C₅H₉ClO₂ Melting Pt: 18 °C Density: 1,17 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004415 CAS Number: 1119-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...