2-Methyl-6-nitro-1H-benzimidazole 98%
Supplier: Apollo Scientific
OR911105-10G
OR911105-1G
OR911105-5G
APOSOR911105-10GEA
434
GBP
APOSOR911105-10G
APOSOR911105-1G
APOSOR911105-5G
2-Methyl-6-nitro-1H-benzimidazole 98%
2-Methyl-6-nitro-1H-benzimidazole
Formula:
C₈H₇N₃O₂ MW: 177,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00748168 CAS Number: 1792-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...