6-Cyano-1-ethylbenzimidazole 98%
Supplier: Apollo Scientific
OR910149-1G
OR910149-25G
OR910149-5G
APOSOR910149-1GEA
132
GBP
APOSOR910149-1G
APOSOR910149-25G
APOSOR910149-5G
6-Cyano-1-ethylbenzimidazole 98%
6-Cyano-1-ethylbenzimidazole
Formula:
C₁₀H₉N₃ Storage Temperature: Ambient |
MDL Number:
MFCD13195705 CAS Number: 1215205-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...