1-(4-Nitrophenyl)-1H-pyrazole 98%

Supplier: Apollo Scientific

OR910032-10G OR910032-25G
APOSOR910032-10GEA 185 GBP
APOSOR910032-10G APOSOR910032-25G
1-(4-Nitrophenyl)-1H-pyrazole 98%
1-(4-Nitrophenyl)-1H-pyrazole
Formula: C₉H₇N₃O₂
MW: 189,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00187828
CAS Number: 3463-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR