5-Bromo-7-chloroquinoxaline 97%
Supplier: Apollo Scientific
|
Danger
|
OR909331-1G
OR909331-250MG
APOSOR909331-1GEA
356
GBP
APOSOR909331-1G
APOSOR909331-250MG
5-Bromo-7-chloroquinoxaline 97%
5-Bromo-7-chloroquinoxaline
Formula:
C₈H₄BrClN₂ MW: 243,487 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15143560 CAS Number: 1215205-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...