3,6-Dimethoxypyridazine 98%

Supplier: Apollo Scientific

OR909190-1G OR909190-5G
APOSOR909190-1GEA 151 GBP
APOSOR909190-1G APOSOR909190-5G
3,6-Dimethoxypyridazine 98%
3,6-Dimethoxypyridazine
Formula: C₆H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00234224
CAS Number: 4603-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR