6-Methylquinoline 98%
Supplier: Apollo Scientific
OR908976-100G
OR908976-25G
APOSOR908976-100GEA
243
GBP
APOSOR908976-100G
APOSOR908976-25G
6-Methylquinoline 98%
6-Methylquinoline
Formula:
C₁₀H₉N MW: 143,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006804 CAS Number: 91-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...