[4-(1-Ethoxyethyl)phenyl]boronic acid 97%

Supplier: Apollo Scientific

OR908323-1G OR908323-250MG OR908323-5G
APOSOR908323-1GEA 197 GBP
APOSOR908323-1G APOSOR908323-250MG APOSOR908323-5G
[4-(1-Ethoxyethyl)phenyl]boronic acid 97%
[4-(1-Ethoxyethyl)phenyl]boronic acid
Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD08059887
CAS Number: 1287753-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR