Phenoxyacetic acid 98%

Supplier: Apollo Scientific

Synonyms: 2-Phenoxyacetic acid

OR908179-100G OR908179-1KG OR908179-500G
APOSOR908179-100GEA 48.6 GBP
APOSOR908179-100G APOSOR908179-1KG APOSOR908179-500G
Phenoxyacetic acid 98%
Phenoxyacetic acid
Formula: C₈H₈O₃
MW: 152,15 g/mol
Boiling Pt: 285 °C/760mmHg
Melting Pt: 98…101 °C
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00004296
CAS Number: 122-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR