Methyl-4-bromobutyrate 98%

Supplier: Apollo Scientific

OR907686-100G OR907686-25G
APOSOR907686-100GEA 33.6 GBP
APOSOR907686-100G APOSOR907686-25G
Methyl-4-bromobutyrate 98%
Methyl 4-bromobutyrate
Formula: C₅H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD00041482
CAS Number: 4897-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR