7-Chloro-1H-indole-2-carboxylic acid 97%
Supplier: Apollo Scientific
OR906974-1G
OR906974-5G
APOSOR906974-1GEA
171
GBP
APOSOR906974-1G
APOSOR906974-5G
7-Chloro-1H-indole-2-carboxylic acid 97%
7-Chloro-1H-indole-2-carboxylic acid
Formula:
C₉H₆ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD02664428 CAS Number: 28899-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...