Ethyl (4-bromobenzoyl)acetate 98%

Supplier: Apollo Scientific
Warning

Synonyms: ethyl 3-(4-bromophenyl)-3-oxopropanoate, 3-(4-bromophenyl)-3-oxo-propionic acid ethyl ester, Benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester

OR906911-25G OR906911-5G
APOSOR906911-25GEA 205 GBP
APOSOR906911-25G APOSOR906911-5G
Ethyl (4-bromobenzoyl)acetate 98%
Ethyl (4-bromobenzoyl)acetate
Formula: C₁₁H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00231243
CAS Number: 26510-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR