2,5-Dimethoxyphenol 95%
Supplier: Apollo Scientific
OR905695-1G
OR905695-5G
APOSOR905695-1GEA
104
GBP
APOSOR905695-1G
APOSOR905695-5G
2,5-Dimethoxyphenol 95%
2,5-Dimethoxyphenol
Formula:
C₈H₁₀O₃ MW: 154,1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00094237 CAS Number: 18113-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...