2-Amino-3-methoxybenzonitrile 96%
Supplier: Apollo Scientific
OR905652-1G
OR905652-250MG
APOSOR905652-1GEA
178
GBP
APOSOR905652-1G
APOSOR905652-250MG
2-Amino-3-methoxybenzonitrile 96%
2-Amino-3-methoxybenzonitrile
Formula:
C₈H₈N₂O MW: 148,1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09744022 CAS Number: 148932-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...