4-Amino-N-methylphthalimide 98%
Supplier: Apollo Scientific
OR905429-100G
OR905429-25G
OR905429-5G
APOSOR905429-100GEA
728
GBP
APOSOR905429-100G
APOSOR905429-25G
APOSOR905429-5G
4-Amino-N-methylphthalimide 98%
4-Amino-N-methylphthalimide
Formula:
C₉H₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00012081 CAS Number: 2307-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...