2-Ethyl-1H-benzo[d]imidazole 95%
Supplier: Apollo Scientific
Synonyms:
2-Ethyl-1H-benzimidazole
OR905351-1G
OR905351-5G
APOSOR905351-1GEA
44.6
GBP
APOSOR905351-1G
APOSOR905351-5G
2-Ethyl-1H-benzo[d]imidazole 95%
2-Ethyl-1H-benzo[d]imidazole
Formula:
C₉H₁₀N₂ MW: 146,1900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00022684 CAS Number: 1848-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...