4-Indanol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2,3-Dihydro-1H-inden-4-ol

OR904926-1G OR904926-250MG OR904926-5G
APOSOR904926-1GEA 60.3 GBP
APOSOR904926-1G APOSOR904926-250MG APOSOR904926-5G
4-Indanol 97%
4-Indanol
Formula: C₉H₁₀O
MW: 134,18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00053431
CAS Number: 1641-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR