3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one 95%

Supplier: Apollo Scientific

Synonyms: 3,5-Dichloro-2,6-dimethylpyridin-4-ol, Clopidol, 3,5-Dichloro-2,6-dimethyl-4-hydroxypyridine

OR904485-10G OR904485-1G OR904485-5G
APOSOR904485-10GEA 408 GBP
APOSOR904485-10G APOSOR904485-1G APOSOR904485-5G
3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one 95%
3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one
Formula: C₇H₇Cl₂NO
MW: 192,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22123390
CAS Number: 2971-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR