1-Furfurylpyrrole 95%

Supplier: Apollo Scientific

Synonyms: 1-(Furan-2-ylmethyl)-1H-pyrrole

OR904283-25G OR904283-5G
APOSOR904283-25GEA 139 GBP
APOSOR904283-25G APOSOR904283-5G
1-Furfurylpyrrole 95%
1-Furfurylpyrrole
Formula: C₉H₉NO
MW: 147,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010080
CAS Number: 1438-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR