2-(4-Methylphenyl)azepane 95%

Supplier: Apollo Scientific

OR904224-1G OR904224-250MG
APOSOR904224-1GEA 617 GBP
APOSOR904224-1G APOSOR904224-250MG
2-(4-Methylphenyl)azepane 95%
2-(4-Methylphenyl)azepane
Formula: C₁₃H₁₉N
Storage Temperature: Ambient
MDL Number: MFCD02663755
CAS Number: 168890-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR