(R)-1-(2-Chlorophenyl)ethanol 97%

Supplier: Apollo Scientific

OR903871-10G OR903871-1G OR903871-5G
APOSOR903871-10GEA 367 GBP
APOSOR903871-10G APOSOR903871-1G APOSOR903871-5G
(R)-1-(2-Chlorophenyl)ethanol 97%
(R)-1-(2-Chlorophenyl)ethanol
Formula: C₈H₉ClO
MW: 156,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659510
CAS Number: 120466-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR