2-Chloro-N-propylacetamide 98%

Supplier: Apollo Scientific

OR903706-10G OR903706-1G OR903706-5G
APOSOR903706-10GEA 446 GBP
APOSOR903706-10G APOSOR903706-1G APOSOR903706-5G
2-Chloro-N-propylacetamide 98%
2-Chloro-N-propylacetamide
Formula: C₅H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD01355490
CAS Number: 13916-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR