2-(3-Methylphenyl)azepane 95%

Supplier: Apollo Scientific

OR903599-1G OR903599-250MG
APOSOR903599-1GEA 617 GBP
APOSOR903599-1G APOSOR903599-250MG
2-(3-Methylphenyl)azepane 95%
2-(3-Methylphenyl)azepane
Formula: C₁₃H₁₉N
Storage Temperature: Ambient
MDL Number: MFCD02663746
CAS Number: 383129-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR