1,2,3,6-Tetrahydropyridine
Supplier: Apollo Scientific
|
Danger
|
OR9035-1G
OR9035-5G
APOSOR9035-1GEA
145
GBP
APOSOR9035-1G
APOSOR9035-5G
1,2,3,6-Tetrahydropyridine
1,2,3,6-Tetrahydropyridine
Formula:
C₅H₉N MW: 83,13 g/mol Boiling Pt: 108 °C Melting Pt: –48 °C Density: 0,911 g/cm³ (25 °C) Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005949 CAS Number: 694-05-3 EINECS: 211-766-2 UN: 2410 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...