5-(Hydroxymethyl)pyridin-2-ol 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Hydroxy-5-hydroxymethylpyridine

OR902374-1G OR902374-250MG
APOSOR902374-1GEA 315 GBP
APOSOR902374-1G APOSOR902374-250MG
5-(Hydroxymethyl)pyridin-2-ol 98%
5-(Hydroxymethyl)pyridin-2-ol
Formula: C₆H₇NO₂
MW: 125,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263739
CAS Number: 109205-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR