2,5-Xylylacetic acid 95%

Supplier: Apollo Scientific

Synonyms: 2-(2,5-Dimethylphenyl)acetic acid, (2,5-Dimethylphenyl)acetic acid

OR902270-100G OR902270-25G
APOSOR902270-100GEA 276 GBP
APOSOR902270-100G APOSOR902270-25G
2,5-Xylylacetic acid 95%
2,5-Xylylacetic acid
Formula: (C₆H₃(CH₃)₂)CH₂COOH
MW: 164,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014369
CAS Number: 13612-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR