1-(5-Methylpyridin-2-yl)-1H-pyrrole-2-carbaldehyde 95%

Supplier: Apollo Scientific

OR902246-1G OR902246-250MG
APOSOR902246-1GEA 617 GBP
APOSOR902246-1G APOSOR902246-250MG
1-(5-Methylpyridin-2-yl)-1H-pyrrole-2-carbaldehyde 95%
1-(5-Methylpyridin-2-yl)-1H-pyrrole-2-carbaldehyde
Formula: C₁₁H₁₀N₂O
MW: 186,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02665160
CAS Number: 142044-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR