3-Bromopropionic acid 98%

Supplier: Apollo Scientific
Danger

Synonyms: 3-Bromopropanoic acid

OR8993-100G OR8993-500G
APOSOR8993-100GEA 103 GBP
APOSOR8993-100G APOSOR8993-500G
3-Bromopropionic acid 98%
3-Bromopropionic acid
Formula: BrH₂CCH₂COOH
MW: 152,98 g/mol
Boiling Pt: 142 °C
Melting Pt: 63 °C
Density: 1,48 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00002763
CAS Number: 590-92-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR